Learn More
ARNT1/HIF-1Β Mouse anti-Human, Mouse, Clone: 4G9, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05704
Catalog No. 05-704-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect ARNT1/HIF-1 beta with Anti-ARNT1/HIF-1β Antibody, clone 4G9 (Mouse Monoclonal Antibody), that has been shown to work in immunoprecipitation, western blot, immunocytochemistry.
Specifications
Specifications
ARNT1/HIF-1β
4G9
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Purified
Murine
IgG
200μg
Primary
Human, Murine
Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
NM_001668
ARNT, HIF-1beta, HIF1beta, HIF1B, HIF1BETA, TANGO
Protein A fusion protein corresponding to residues 399-777 of human aryl hydrocarbon receptor nuclear translocator (ARNT)
Protein G chromatography
−20°C for 24 months.
Monoclonal