Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Arrestin 3, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP137003

Catalog No. NBP137003


Add to cart

Description

Description

Arrestin 3 Polyclonal antibody specifically detects Arrestin 3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications

Specifications

Arrestin 3
0.5 mg/mL
Immunohistochemistry 2.5 ug/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 2.5 ug/mL, Peptide ELISA Detection limit 1:4000
NP_004303.1
arrestin 3, retinal (X-arrestin), arrestin 4, arrestin-C, ARRXcArr, CAR, C-arrestin, Cone arrestin, Retinal cone arrestin-3, X-arrestin
Goat
IgG
0.1 mg
Cell Biology, Cellular Markers, GPCR, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Virology Bacteria and Parasites, Vision
Primary
407
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
ARR3
Arrestin 3 Antibody is made to a peptide with sequence C-RKGEEESQKAVE corresponding to C-Terminus according to NP_004303.2.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is specific to the C Terminus.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only