Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arylsulfatase A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522337

Catalog No. PIPA522337


Add to cart

Description

Description

PA5-22337 targets ARSA in ICC, IF, IHC (P), and WB applications and shows reactivity with Human and mouse samples. Predicted to react with Rat, Xenopus laevis, Zebrafish, Bovine, and Xenopus tropicalis based on sequence homology. The PA5-22337 immunogen is recombit protein fragment contain a sequence corresponding to a region within amino acids 45 and 279 (O43681) of ASNA1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

Arylsulfatase A
Polyclonal
Unconjugated
ARSA
Liquid
ARSA
Recombit protein fragment contain a sequence corresponding to a region within amino acids 45 and 279 (O43681) of ASNA1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.76 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
P15289, P50428
ARSA-I, ARSA1, ASNA-I, GET3, TRC40, hASNA-I, ATPase ASNA1, arsenical pump-driving ATPase, arsenite-stimulated ATPase, golgi to ER traffic 3 homolog, transmembrane domain recognition complex 40 kDa ATPase subunit, transmembrane domain recognition complex, 40kDa
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
11883, 410
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit