Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADP-ribosyltransferase 3 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ART3 protein.

Manufacturer:  Abnova Corporation H00000419B01

Catalog No. 89-004-942


Add to cart

Description

Description

ADP-ribosylation is a reversible posttranslational modification used to regulate protein function. ADP-ribosyltransferases, such as ART3, transfer ADP-ribose from NAD+ to the target protein, and ADP-ribosylhydrolases (see PARG; MIM 603501) reverse the reaction (Glowacki et al., 2002 [PubMed 12070318]).[supplied by OMIM

Sequence: MKTGHFEIVTMLLATMILVDIFQVKAEVLDMADNAFDDEYLKCTDRMEIKYVPQLLKEEKASHQQLDTVWENAKAKWAARKTQIFLPMNFKDNHGIALMAYISEAQEQTPFYHLFSEAVKMAGQSREDYIYGFQFKAFHFYLTRALQLLRKPCEASSKTVVYRTSQGTSFTFGGLNQARFGHFTLAYSAKPQAANDQLTVLSIYTCLGVDIENFLDKESERITLIPLNEVFQVSQEGAGNNLILQSINKTCSHYECAFLGGLKTENCIENLEYFQPIYVYNPGEKNQKLEDHSEKNWKLEDHGEKNQKLEDHGVKILEPTQIPEDKSQGNINNPTPGPVPVPGPKSHPSASSGKLLLPQFGMVIILISVSAINLFVAL
Specifications

Specifications

ADP-ribosyltransferase 3
Unconjugated
No additive
NM_001179.2
FLJ26404
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
419
Western Blot
Mouse polyclonal antibody raised against a full-length human ART3 protein.
ART3
Serum
ART3
ART3 (NP_001170.2, 1 a.a. ~ 378 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit