Learn More
ADP-ribosyltransferase 5, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ART5 protein.
Manufacturer: Abnova Corporation H00116969B01
Description
The protein encoded by this gene belongs to the ARG-specific ADP-ribosyltransferase family. Proteins in this family regulate the function of target proteins by attaching ADP-ribose to specific amino acid residues in their target proteins. The mouse homolog lacks a glycosylphosphatidylinositol-anchor signal sequence and is predicted to be a secretory enzyme. Transcript variants with different 5' UTRs, but encoding the same protein have been found for this gene. [provided by RefSeq
Sequence: MALAALMIALGSLGLHTWQAQAVPTILPLGLAPDTFDDTYVGCVEEMEEKAAPLLKEEMAHHALLRESWEAAQETWEDKRRGLTLPPGFKAQNGIAIMVYTNSSNTLYWELNQAVRTGGGSRELYMRHFPFKALHFYLIRALQLLRGSGGCSRGPGEVVFRGVGSLRFEPKRLGDSVRLGQFASSSLDKAVAHRFGNATLFSLTTCFGAPIQAFSVFPKEREVLIPPHEVFLVTRFSQDGAQSLVTLWSYNQTCSHFNCAYLGGEKRRGCVSAPGALGTGDLHMTKRHLQQPSpecifications
ADP-ribosyltransferase 5 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human ART5 protein. | |
ART5 | |
MGC22848 | |
Mouse | |
50 μL | |
Yes | |
116969 | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
BC014577 | |
ART5 | |
ART5 (AAH14577, 1 a.a. ∼ 292 a.a) full-length human protein. | |
RUO | |
Primary | |
Human |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.