Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Artemis Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527112

Catalog No. PIPA527112


Add to cart

Description

Description

PA5-27112 targets Artemis in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-27112 immunogen is recombit fragment corresponding to a region within amino acids 306 and 692 of Human Artemis. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a nuclear protein that is involved in V(D)J recombination and DNA repair. The protein has single-strand-specific 5 ft.-3 ft. exonuclease activity; it also exhibits endonuclease activity on 5 ft. and 3 ft. overhangs and hairpins when complexed with protein kinase, DNA-activated, catalytic polypeptide. Mutations in this gene cause Athabascan-type severe combined immunodeficiency (SCIDA).
Specifications

Specifications

Artemis
Polyclonal
Unconjugated
DCLRE1C
Liquid
DCLRE1C
Recombit fragment corresponding to a region within amino acids 306 and 692 of Human Artemis
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ChIP assay, Immunohistochemistry (Paraffin), Western Blot
0.78 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q96SD1
A-SCID, DCLREC1C, RS-SCID, SCIDA, SNM1C, hSNM1C, DNA cross-link repair 1C (PSO2 homolog, S. cerevisiae), DNA cross-link repair 1C protein, PSO2 homolog, SNM1 homolog C, SNM1-like protein, protein artemis, severe combined immunodeficiency, type a (Athabascan)
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
64421
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit