Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARTS1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515021

Catalog No. PIPA515021

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Aminopeptidases play a role in the metabolism of several peptides that may be involved in blood pressure and the pathogenesis of essential hypertension. Adipocyte-derived leucine aminopeptidase (ALAP) is a member of the M1 family of zinc metallopeptidases.
Specifications

Specifications

ARTS1
Polyclonal
PBS with 0.09% sodium azide
Q9NZ08
Endoplasmic reticulum aminopeptidase 1; Adipocyte-derived leucine aminopeptidase; A-LAP; ARTS-1; Aminopeptidase PILS; Puromycin-insensitive leucyl-specific aminopeptidase; PILS-AP; Type 1 tumor necrosis factor receptor shedding aminopeptidase regulator; ERAP1; APPILS; ARTS1; KIAA0525
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51752
Western Blot
Unconjugated
ERAP1
Liquid
ERAP1
KLH conjugated synthetic peptide between 451-480 amino acids from the central region of human ARTS1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit