Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARV1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524151

Catalog No. PIPA524151

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with rat based on sequence homology.

May act as a mediator of sterol homeostasis.
Specifications

Specifications

ARV1
Polyclonal
PBS with 0.09% sodium azide
Q9H2C2, Q9D0U9
hARV1, protein ARV1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64801, 68865
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
ARV1
Liquid
ARV1
KLH conjugated synthetic peptide between 23-51 amino acids from the N-terminal region of human ARV1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit