Learn More
AHR Goat anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Manufacturer:  Invitrogen PA146110
Catalog No. PA146110
Add to cart
Description
Description
Suggested positive control: antigen standard for AHR (transient overexpression lysate). This antibody is not recommended for Western Blot applications.
AHR (Ah Receptor) belongs to a family of proteins comprised of its dimerization partner ARNT (HIF-1 Beta) and the Drosophila proteins PER and SIM. AHR contains an N-terminal sequence of approximately 200 amino acids termed the PAS domain. AHR, found in a variety of tissues, binds to a specific DNA enhancer sequence and initiates transcription of the mRNA for the cytochrome P-450 (CYPIA1) gene. The gene for AHR encodes a ligand-activated transcription factor involved in the regulation of biological responses to planar aromatic hydrocarbons. AHR has been shown to regulate xenobiotic-metabolizing enzymes such as cytochrome P450, and its ligands included a variety of aromatic hydrocarbons. AHR is a ligand-activated helix/loop/helix transcription factor found in a variety of vertebrate species. The known ligands for AHR are foreign planar aromatic compounds, such as polycyclic aromatic compounds and halogenated aromatic compounds such as 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD). Unlike the steroid/thyroid hormone receptors, there is no known physiological ligand for AHR. Studies indicate that in non-ligand activated cells, AHR is found complexed with HSP90 predomitly in the cytoplasm. Upon binding to an agonist, the ligand-activated AhR is believed to transform to a nuclear, DNA binding form, and this transformation process appears to involve dissociation of HSP90 from AhR followed by formation of a heterodimer with AhR nuclear translocator protein (Arnt). Diseases associated with AHR include eosinophilic fasciitis and seborrheic dermatitis.
Specifications
Specifications
AHR
Polyclonal
whole serum with no preservative
P35869, P30561, P41738
bHLHe76
Goat
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ChIP assay, Gel Shift, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Unconjugated
AHR
Liquid
AHR
N-terminal sequence of Aryl hydrocarbon Receptor purified from C57BL/6J mice
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11622, 196, 25690