Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ASB11, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB10056640SS

 View more versions of this product

Catalog No. NB10056640T


Add to cart

Description

Description

ASB11 Polyclonal specifically detects ASB11 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ASB11
0.5 mg/mL
Western Blot 1:500-1:1000
Purified
ASB11
This antibody was developed against synthetic peptides corresponding to amino acids 40-51 and 250-265 of human Asb-11.
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Unconjugated
PBS containing 0.1 mg/ml BSA. with 0.02% Sodium Azide
ankyrin repeat and SOCS box containing 11, ankyrin repeat and SOCS box protein 11, ankyrin repeat and SOCS box-containing 11, ankyrin repeat domain-containing SOCS box protein ASB11, ASB-11, DKFZp779E2460, MGC119168, MGC119169
Rabbit
IgG
0.05 ml
Signal Transduction
Primary
140456
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only