Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASB17 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524312

Catalog No. PIPA524312


Add to cart

Description

Description

This antibody is predicted to react with non-human primate based on sequence homology.

May be a substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
Specifications

Specifications

ASB17
Polyclonal
PBS with 0.09% sodium azide
Q8WXJ9, Q8VHP9
Asb-17, ankyrin repeat and SOCS box protein 17, ankyrin repeat and SOCS box-containing 17
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
127247, 66772
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ASB17
Liquid
ASB17
KLH conjugated synthetic peptide between 181-209 amino acids from the C-terminal region of human ASB17
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit