Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ASB2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB10056638

 View more versions of this product

Catalog No. NB10056638


Add to cart

Description

Description

ASB2 Polyclonal antibody specifically detects ASB2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ASB2
0.5 mg/mL
Western Blot 1:500-1:1000
Q96Q27
ankyrin repeat and SOCS box containing 2, ankyrin repeat and SOCS box protein 2, ankyrin repeat and SOCS box-containing protein 2a, ASB-2ankyrin repeat and SOCS box-containing 2, MGC40044
Rabbit
IgG
0.2 ml
Signal Transduction
Primary
51676
Human
Western Blot
Unconjugated
Whole antisera with PBS and 0.05% BSA with 0.05% Sodium Azide
Antisera
ASB2
This polyclonal antibody was raised against synthetic peptides containing amino acids 75-93 and 348-368 of human Asb-2 protein. Human spleen lysate can be used as a positive control. It should cross react with mouse and rat Asb-2, based on amino acid sequences used.
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Cross reacts with Human. Predicted to react with Mouse and Rat.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only