Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASB2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141304

Catalog No. 01-675-644


Add to cart

Description

Description

This antibody may cross react with mouse and rat Asb-2, based on sequence homology. Suggested positive control: human spleen protein, human lung or spleen, human lung protein.

The ankyrin repeat and SOCS box-containing (Asb) protein family is comprised of members which each contain “SOCS boxes” but differ from other families of protein in the motifs they contain upstream of the SOCS box(1). SOCS boxes are carboxy-terminal regions of homology found in the suppressor of cytokine signaling family of proteins. The box region is thought to be the point of interaction between SOCS proteins and E3 ubiquitin ligases. Asb-2 has been found to induce growth inhibition and chromatin condensation in myeloid leukemia cells(2).
Specifications

Specifications

ASB2
Polyclonal
whole serum diluted in PBS with 0.05% BSA and 0.05% sodium azide
Q96Q27
ASB-2, MGC40044
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Western Blot
Unconjugated
ASB2
Liquid
ASB2
Synthetic peptides corresponding to residues 75-93 and 348-368 of human Asb-2.
200 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
51676
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit