Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASB3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA129823

Catalog No. PIPA129823


Add to cart

Description

Description

PA1-29823 detects ASB3 from human samples. PA1-29823 has been successfully used in Western blot applications. The PA1-29823 immunogen is: Synthetic peptide: DSSENYIKMK corresponding to amino acids 66 to 75 of Human ASB3 and IRKGANKECQDDFGI corresponding to amino acids 166 to 180 of Human ASB3.

The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene but some of the full length sequences are not known.
Specifications

Specifications

ASB3
Polyclonal
whole serum diluted in PBS with 0.01mg/mL BSA and 0.02% sodium azide
Q9Y575
ASB-3, FLJ10123, FLJ10421, MGC12531, MGC132002, MGC996
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
51130
Western Blot
Unconjugated
ASB3
Liquid
ASB3
synthetic peptide DSSENYIKMK corresponding to amino acids 66 to 75 of Human ASB3 and IRKGANKECQDDFGI corresponding to amino acids 166 to 180 of Human ASB3.
Protein G
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit