Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ASC/TMS1, DyLight 650, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178978C

Catalog No. NBP178978C


Add to cart

Description

Description

ASC/TMS1 Polyclonal antibody specifically detects ASC/TMS1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Western Blot (Negative).
Specifications

Specifications

ASC/TMS1
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
PYCARD
A synthetic peptide made to a C-terminal portion of the human ASC/TMS1 protein (between residues 145-195) [Uniprot: Q9ULZ3]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:40-1:100, Immunohistochemistry-Paraffin 1:200, Western Blot (Negative)
Q9ULZ3
ASCMGC10332, CARD5TMS, caspase recruitment domain protein 5, Caspase recruitment domain-containing protein 5, hASC, PYD and CARD domain containing, PYD and CARD domain-containing protein, Target of methylation-induced silencing 1, TMS-1, TMS1apoptosis-associated speck-like protein containing a CARD
Rabbit
IgG
0.1 ml
Apoptosis, Cancer, Cell Biology
Primary
29108
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only