Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASC/TMS1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178978H
Description
ASC/TMS1 Polyclonal specifically detects ASC/TMS1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative).Specifications
ASC/TMS1 | |
Polyclonal | |
Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200, Western Blot (Negative) | |
Q9ULZ3 | |
PYCARD | |
This ASC/TMS1 Antibody was developed against a synthetic peptide made to a C-terminal portion of the human ASC/TMS1 protein (between residues 145-195) [Uniprot: Q9ULZ3] | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Inflammation, Innate Immunity | |
29108 | |
Store at 4C in the dark. |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
HRP | |
PBS with No Preservative | |
ASCMGC10332, CARD5TMS, caspase recruitment domain protein 5, Caspase recruitment domain-containing protein 5, hASC, PYD and CARD domain containing, PYD and CARD domain-containing protein, Target of methylation-induced silencing 1, TMS-1, TMS1apoptosis-associated speck-like protein containing a CARD | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction