Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ASC/TMS1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178977SS

 View more versions of this product

Catalog No. NBP178977SS


Add to cart

Description

Description

ASC/TMS1 Polyclonal antibody specifically detects ASC/TMS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

ASC/TMS1
1.0 mg/mL
Western Blot 2.0 - 4.0 ug/ml, Simple Western 1:1000, Flow Cytometry, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:40-1:100, Immunoprecipitation, Immunohistochemistry-Paraffin 1:400, Immunomicroscopy
Q9ULZ3
ASCMGC10332, CARD5TMS, caspase recruitment domain protein 5, Caspase recruitment domain-containing protein 5, hASC, PYD and CARD domain containing, PYD and CARD domain-containing protein, Target of methylation-induced silencing 1, TMS-1, TMS1apoptosis-associated speck-like protein containing a CARD
Rabbit
IgG
0.025 ml
Apoptosis, Cancer, Cell Biology, Inflammation, Innate Immunity
Primary
29108
Human, Mouse, Rat
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunomicroscopy
Unconjugated
PBS and 30% Glycerol with 0.05% Sodium Azide
Affinity Purified
PYCARD
This ASC/TMS1 Antibody was developed against a synthetic peptide made to an N-terminal portion of the human ASC/TMS1 protein (between residues 1-50) [Uniprot: Q9ULZ3]
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Reactivity with Rat reported in PMID 24464748 .
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only