Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ASC/TMS1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide corresponding to amino acids 83-102 (GQLQAATHQGSGAAPAGIQA) of human ASC/TMS1 was used as immunogen; GenBank no. NP_037390.2, which is also referred to as ASC/TMS1 isoform a. Isoform a is a 195 amino acid protein. Amino acids 85-102 of the peptide immunogen immunogen are 100% consevered in ASC/TMS1 isoform c, GenBank no. NP_660184.1. Isoform c is a 135 amino acid protein.
Antigen ASC/TMS1
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056075T
View Documents
Novus Biologicals
NB10056075SS
0.025 ml This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
NB10056075
View Documents
Novus Biologicals
NB10056075
0.05 ml This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

ASC/TMS1 Polyclonal antibody specifically detects ASC/TMS1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications

Specifications

Rabbit
A synthetic peptide corresponding to amino acids 83-102 (GQLQAATHQGSGAAPAGIQA) of human ASC/TMS1 was used as immunogen; GenBank no. NP_037390.2, which is also referred to as ASC/TMS1 isoform a. Isoform a is a 195 amino acid protein. Amino acids 85-102 of the peptide immunogen immunogen are 100% consevered in ASC/TMS1 isoform c, GenBank no. NP_660184.1. Isoform c is a 135 amino acid protein.
Polyclonal
Primary
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
29108
IgG
ASC/TMS1
Antisera
RUO
ASC
Whole antisera with 0.05% Sodium Azide
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit