Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASCL1 Rat anti-Mouse, Clone: 290622, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA523994

Catalog No. PIMA523994


Add to cart

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

ASCL1 (also known as ASH1) is a basic helix-loop-helix transcription factor that is required for early development of the nervous system. Expressed in fetal brain, ASCL1 is essential for the proper development of autonomic neurons and for the survival of subsets of autonomic neurons. ASCL1 interaction with MEF-2A may regulate the expression of specific genes that are critical for the formation of distinct neuronal circuits within the central nervous system. The high level of ASCL1 expression in neuroendocrine tumors, such as medullary thyroid cancer, small cell lung cancer and lung cancer with neuroendocrine features may provide a useful marker for cancers with neuroendocrine features. Mapping to human chromosome 12, the ASCL1 gene contains a trinucleotide repeat region, making this locus a candidate for inherited disease.
Specifications

Specifications

ASCL1
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q02067
Achaete-scute homolog 1, ASH-1, hASH1, Class A basic helix-loop-helix protein 46, bHLHa46, ASCL1, ASH1, BHLHA46, HASH1, Mash-1, Mash1
Rat
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
17172
Immunocytochemistry, Immunofluorescence, Western Blot
290622
Unconjugated
Ascl1
Lyophilized
ASCL1
E. coli-derived recombit mouse ASCL1/Mash1 His74-Asp173 (Gln160Glu)
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit