Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASGR2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124416

Catalog No. PA124416


Add to cart

Description

Description

PA1-24416 detects all three isoforms of asialoglycoprotein receptor 2 in human samples. PA1-24416 has been successfully used in Western blot and immunohistochemistry (FFPE) procedures. The PA1-24416 immunogen is a synthetic peptide corresponding to a region of human ASGR2. For use in Western Blot, it is suggested that PA1-24416 is diluted in 5% skim milk/PBS buffer.

ASGR2 is a cell surface receptor that binds to galactose-terminated glycoproteins. It transports these glycoproteins via a series of membrane vesicles and tubules to an acidic-sorting organelle where the receptor and ligand dissociates. Then the receptor is recycled back to the cell surface. There are four alternatively spliced transcript variants of this gene. This gene has multiple polyadenylation sites.
Specifications

Specifications

ASGR2
Polyclonal
Unconjugated
ASGR2
Liquid
ASGR2
Synthetic peptide corresponding to a region of human ASGR2.
Protein A
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide; pH 7.2
P07307
ASGR2
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
433
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit