Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASMT Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524721

Catalog No. PIPA524721


Add to cart

Description

Description

ASMT belongs to the methyltransferase superfamily, and is located in the pseudoautosomal region (PAR) at the end of the short arms of the X and Y chromosomes. The encoded enzyme catalyzes the final reaction in the synthesis of melatonin, and is abundant in the pineal gland.
Specifications

Specifications

ASMT
Polyclonal
PBS with 0.09% sodium azide
P46597
ASMTY, HIOMT, HIOMTY, acetylserotonin N-methyltransferase, acetylserotonin methyltransferase (Y chromosome), hydroxyindole O-methyltransferase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
438
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ASMT
Liquid
ASMT
KLH conjugated synthetic peptide between 212-241 amino acids from the central region of human ASMT
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit