Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aspartate Aminotransferase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
Supplier: Novus Biologicals NBP154778
Description
Aspartate Aminotransferase Polyclonal specifically detects Aspartate Aminotransferase in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Aspartate Aminotransferase | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
aspartate aminotransferase, cytoplasmic, EC 2.6.1.1, GIG18, Glutamate oxaloacetate transaminase 1, glutamic-oxaloacetic transaminase 1, soluble (aspartate aminotransferase 1), growth-inhibiting protein 18, Transaminase A | |
Rabbit | |
46 kDa | |
100 μL | |
Lipid and Metabolism | |
2805 | |
Human, Mouse, Rat, Bovine, Equine, Guinea Pig, Goat, Rabbit, Yeast, Zebrafish | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
P17174 | |
GOT1 | |
Synthetic peptides corresponding to GOT1(glutamic-oxaloacetic transaminase 1, soluble (aspartate aminotransferase 1)) The peptide sequence was selected from the N terminal of GOT1 (NP_002070). Peptide sequence MAPPSVFAEVPQAQPVLVFKLTADFREDPDPRKVNLGVGAYRTDDCHPWV The peptide sequence for this immunogen was taken from within the described region. | |
Protein A purified | |
RUO | |
Primary | |
Centrifuge at 12,000 x g for 20 seconds. Reconstitute with 0.1 ml distilled water to a final concentration of 1.0 mg/ml in PBS buffer. Vortex followed by centrifuge again to pellet the solution. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction