Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aspergillus niger Glucose Oxidase, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA185473

Catalog No. PA185473

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Aspergillus species are highly aerobic and are found in almost all oxygen-rich environments, where they commonly grow as molds on the surface of a substrate, as a result of the high oxygen tension. Some Aspergillus species cause serious disease in humans

Specifications

Specifications

Aspergillus niger Glucose Oxidase
Polyclonal
whole serum with 0.09% sodium azide
A. niger Glucose Oxidase, Aspergillus niger GOx, A. niger GOx
Rabbit
IgG
RUO
Antibody
Polyclonal
ELISA
Unconjugated
Serum
gox
Native protein.
2mL
Store at 4°C or Store at -20°C if preferred
Primary
Fungi
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit