Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aspergillus Rabbit anti-Fungi, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA17202

Catalog No. PA17202


Add to cart

Description

Description

PA1-7202 detects soluble proteins from common Aspergillus species. PA1-7202 has been successfully used in immunohistochemistry procedures. The PA1-7202 immunogen is soluble extract from A. Fumigatus, A. flavus, A. niger and A. Terreus.

Aspergillus species are highly aerobic and are found in almost all oxygen-rich environments, where they commonly grow as molds on the surface of a substrate, as a result of the high oxygen tension. Some Aspergillus species cause serious disease in humans
Specifications

Specifications

Aspergillus
Polyclonal
Unconjugated
Liquid
Soluble extract from A. Fumigatus, A. flavus, A. niger and A. Terreus.
Purified
RUO
Antibody
Polyclonal
Immunohistochemistry
4-5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Rabbit
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Fungi
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit