Learn More
aspartate beta-hydroxylase, Mouse, Polyclonal Antibody, Abnova
Mouse polyclonal antibody raised against a full-length human ASPH protein.
Manufacturer: Abnova Corporation H00000444B01
Description
This gene is thought to play an important role in calcium homeostasis. The gene is expressed from two promoters and undergoes extensive alternative splicing. The encoded set of proteins share varying amounts of overlap near their N-termini but have substantial variations in their C-terminal domains resulting in distinct functional properties. The longest isoforms (a and f) include a C-terminal Aspartyl/Asparaginyl beta-hydroxylase domain that hydroxylates aspartic acid or asparagine residues in the epidermal growth factor (EGF)-like domains of some proteins, including protein C, coagulation factors VII, IX, and X, and the complement factors C1R and C1S. Other isoforms differ primarily in the C-terminal sequence and lack the hydroxylase domain, and some have been localized to the endoplasmic and sarcoplasmic reticulum. Some of these isoforms are found in complexes with calsequestrin, triadin, and the ryanodine receptor, and have been shown to regulate calcium release from the sarcoplasmic reticulum. Some isoforms have been implicated in metastasis. [provided by RefSeq
Sequence: MAQRKNAKSSGNSSSSGSGSGSTSAGSSSPGARRETKHGGHKNGRKGGLSGTSFFTWFMVIALLGVWTSVAVVWFDLVDYEEVLGKLGIYDADGDGDFDVDDAKVLLGLKERSTSEPAVPPEEAEPHTEPEEQVPVEAEPQNIEDEAKEQIQSLLHEMVHAEHETEHSYHVEETVSQDCNQDMEEMMSEQENPDSSEPVVEDERLHHDTDDVTYQVYEEQAVYEPLENEGIEITEVTAPPEDNPVEDSQVIVEEVSIFPVEEQQEVPPDTSpecifications
aspartate beta-hydroxylase | |
Unconjugated | |
No additive | |
BC025236.1 | |
BAH|CASQ2BP1|HAAH|JCTN|junctin | |
Mouse | |
50 uL | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Primary | |
444 |
Western Blot | |
Mouse polyclonal antibody raised against a full-length human ASPH protein. | |
ASPH | |
Serum | |
ASPH | |
ASPH (AAH25236.1, 1 a.a. ~ 270 a.a) full-length human protein. | |
RUO | |
Y | |
Polyclonal | |
Human |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.