Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Ataxin 1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128734

Catalog No. NBP128734


Add to cart

Description

Description

Ataxin 1 Polyclonal antibody specifically detects Ataxin 1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

Ataxin 1
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 1:10-1:500
NP_000323.2
ataxin 1, ataxin 1), ataxin-1, ATX1spinocerebellar ataxia 1 (olivopontocerebellar ataxia 1, autosomal dominant, SCA1D6S504E, Spinocerebellar ataxia type 1 protein
Rabbit
IgG
0.1 ml
Store at 4C. Do not freeze.
Polyclonal
Based on 100% sequence identity, this antibody is predicted to react with Panda, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, African elephant and Northern white-cheeked gibbon.
Immunoprecipitation, Western Blot
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
Affinity Purified
ATXN1
The immunogen recognized by this antibody maps to a region between residue 766 and 816 of human ataxin-1 using the numbering given in entry NP_000323.2 (GeneID 6310).
Immunogen affinity purified
RUO
Primary
6310
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only