Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATF1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA125328

Catalog No. PIPA125328


Add to cart

Description

Description

PA1-25328 detects ATF1 from human samples. PA1-25328 has been successfully used in Western blot procedures. The PA1-25328 immunogen is a synthetic peptide, conjugated to KLH, corresponding to residues K(6) S T T S E T A P Q P G S A V Q G A(23) of human ATF1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

ATF1
Polyclonal
0.01M PBS with 1% BSA and 15mM sodium azide; pH 7.4
P18846
EWS-ATF1, FUS/ATF-1, TREB36
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
466
Western Blot
Unconjugated
ATF1
Liquid
ATF1
Synthetic peptide, conjugated to KLH, corresponding to residues K(6) S T T S E T A P Q P G S A V Q G A(23) of human ATF1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit