Learn More
ATF3, Mouse, Clone: 8D8, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant ATF3.
Supplier: Abnova Corporation H00000467M02
Description
Activating transcription factor 3 is a member of the mammalian activation transcription factor/cAMP responsive element-binding (CREB) protein family of transcription factors. Multiple transcript variants encoding two different isoforms have been found for this gene. The longer isoform represses rather than activates transcription from promoters with ATF binding elements. The shorter isoform (deltaZip2) lacks the leucine zipper protein-dimerization motif and does not bind to DNA, and it stimulates transcription presumably by sequestering inhibitory co-factors away from the promoter. It is possible that alternative splicing of the ATF3 gene may be physiologically important in the regulation of target genes. [provided by RefSeq
Sequence: MMLQHPGQVSASEVSASAIVPCLSPPGSLVFEDFANLTPFVKEELRFAIQNKHLCHRMSSALESVTVSDRPLGVSITKAEVAPEEDERKKRRRERNKIAAAKCRNKKKEKTECLQKESEKLESVNAELKAQIEELKNEKQHLIYMLNLHRPTCIVRAQNGRTPEDERNLFIQQIKEGTLQSSpecifications
ATF3 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC006322 | |
Mouse | |
Affinity chromatography | |
RUO | |
467 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
ELISA, Western Blot | |
8D8 | |
Mouse monoclonal antibody raised against a full length recombinant ATF3. | |
ATF3 | |
ATF3 | |
ATF3 (AAH06322, 1 a.a. ∼ 181 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.