Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATF6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
$212.00 - $500.50
Specifications
Antigen | ATF6 |
---|---|
Concentration | 0.6 mg/mL |
Dilution | Western Blot 1:100-1:2000, Simple Western 1:25, Immunocytochemistry/Immunofluorescence 1:100 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP175478SS
|
Novus Biologicals
NBP175478SS |
0.025 mL |
Each for $212.00
|
|
|||||
NBP175478
|
Novus Biologicals
NBP175478 |
0.1 mL |
Each for $500.50
|
|
|||||
Description
ATF6 Polyclonal specifically detects ATF6 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.Specifications
ATF6 | |
Western Blot 1:100-1:2000, Simple Western 1:25, Immunocytochemistry/Immunofluorescence 1:100 | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism, Signal Transduction, Unfolded Protein Response | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha | |
ATF6 | |
IgG | |
Affinity Purified |
0.6 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human, Mouse | |
P18850 | |
22926 | |
A genomic peptide made to an internal region of the human ATF6 protein (within residues 200-350). [Swiss-Prot P18850] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title