Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

ATF7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528130

Catalog No. PIPA528130



Description

Description

PA5-28130 targets ATF7 in IF and WB applications and shows reactivity with Human samples. The PA5-28130 immunogen is synthetic peptide corresponding to a region within amino acids 432 and 494 of Human ATF7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

ATF7
Polyclonal
Unconjugated
ATF7
Liquid
ATF7
Synthetic peptide corresponding to a region within amino acids 432 and 494 of Human ATF7
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
0.88 mg/mL
PBS with 10% glycerol and 0.01% thimerosal; pH 7
P17544
ATFA, cyclic AMP-dependent transcription factor ATF-7, transcription factor ATF-A
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
11016
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit