Learn More
ATG12 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSA00023W
Catalog No. PIOSA00023W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 150 μL of distilled water.
APG12 is required for autophagy. APG5 conjugates to ATG12 and associates with isolation membrane to form cup-shaped isolation membrane and autophagosome. The conjugate detaches from the membrane immediately before or after autophagosome formation is completed.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ATG12
Polyclonal
ATG12
APG12, APG12L, FBR93, HAPG12
Rabbit
150 μL
Primary
Human
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
O94817
ATG12
A synthetic peptide from human ATG12 conjugated to an immunogenic carrier protein
RUO
9140
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Recommendations
Recommendations