Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG12 Sheep anti-Mouse, Rat, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Supplier:  Thermo Scientific OSA00025W

Catalog No. PIOSA00025W

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Glycerol (1:1) may be added for additional stability. Reconstitute with 150 μL of distilled water.

Autophagy is a process of bulk protein degradation in which cytoplasmic components, including organelles, are enclosed in double-membrane structures called autophagosomes and delivered to lysosomes or vacuoles for degradation. ATG12 is the human homolog of a yeast protein involved in autophagy [PubMed 9852036].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATG12
Polyclonal
Atg12
APG12, APG12L, FBR93, HAPG12
Sheep
150 μL
Primary
Mouse, Rat
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
Q9CQY1, Q2TBJ5
ATG12
A synthetic peptide from rat, mouse ATG12
RUO
361321, 67526
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit