Learn More
ATG12 Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSA00028W
Catalog No. PIOSA00028W
Add to cart
Description
Description
Glycerol (1:1) may be added for additional stability. Reconstitute with 150 µL of distilled water.
Autophagy is a process of bulk protein degradation in which cytoplasmic components, including organelles, are enclosed in double-membrane structures called autophagosomes and delivered to lysosomes or vacuoles for degradation. ATG12 is the human homolog of a yeast protein involved in autophagy (Mizushima et al., 1998 [PubMed 9852036]).
Specifications
Specifications
ATG12
Polyclonal
whole serum with no preservative
Q9CQY1, Q2TBJ5
APG12, APG12L, FBR93, HAPG12
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse, Rat
Immunohistochemistry, Western Blot
Unconjugated
Atg12
Lyophilized
ATG12
A synthetic peptide from rat ATG12
150 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
361321, 67526