Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG16L1, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA146307

Catalog No. PA146307

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Autophagy is the major intracellular degradation system delivering cytoplasmic components to lysosomes, and it accounts for degradation of most long-lived proteins and some organelles. Cytoplasmic constituents, including organelles, are sequestered into double-membraned autophagosomes, which subsequently fuse with lysosomes. ATG16L1 is a component of a large protein complex essential for autophagy (Mizushima et al., 2003 [PubMed 12665549]).

Suggested positive control: HeLa whole cell lysate.
Specifications

Specifications

ATG16L1
Polyclonal
Unconjugated
ATG16L1
Purified
ATG16L1
Synthetic peptide corresponding to an N-terminal portion within residues 1-100 of the human protein [Swiss-Prot# Q676U5]
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Canine, Human, Murine, Non-human Primate, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blot
0.8mg/mL
PBS with 30% glycerol and 0.1% sodium azide
Q676U5, Q8C0J2
apg16l beta , apg16 autophagy 16like s cerevisiae , wdr30 , wd repeat domain 30 , atg16l anti
Rabbit
IgG
100μL
Store at 4°C short term. For long term storage, Store at -20°C, avoiding freeze/thaw cycles.
Primary
55054
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit