Learn More
ATG3 Sheep anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen OSA00005W
Catalog No. PIOSA00005W
Add to cart
Description
Description
Reconstitute with 150 µL of distilled water. Centrifuge to remove any insoluble material.
Autophagy is a process of bulk degradation of cytoplasmic components by the lysosome or vacuole. Human ATG3 displays the same enzymatic characteristics in vitro as yeast Apg3, a protein-conjugating enzyme essential for autophagy (Tanida et al., 2002 [PubMed 11825910]).
Specifications
Specifications
ATG3
Polyclonal
whole serum with no preservative
Q9NT62, Q9CPX6, Q6AZ50
ATG3, APG3, APG3L, APG3-like, hApg3, protein PC3-96, FLJ22125, MGC15201, DKFZp564M1178
Sheep
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry, Western Blot
Unconjugated
ATG3
Lyophilized
ATG3
A synthetic peptide from human autophagy-related protein 3
150 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
171415, 64422, 67841