Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG4B Rabbit anti-Human, Invitrogen™

Rabbit Recombinant Polyclonal Antibody

Manufacturer:  Invitrogen 710915

Catalog No. PI710915


Add to cart

Description

Description

This antibody is predicted to react with monkey, mouse, rat, guinea pig and bovine based on sequence homology. Recombit rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombit monoclonal antibodies, providing the best of both worlds – the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombit antibody. While functionally the same as a polyclonal antibody – recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets – a recombit rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.

Cysteine protease ATG4B is an enzyme which is coded by ATG4B gene. Autophagy is a critical cellular homeostatic process that controls the turnover of damaged organelles and protein. ATG4B is a deubiquitin-like protease that plays dual roles in the core machinery of autophagy. ATG4B, cleaves the C-terminal amino acid of ATG8 family proteins MAP1LC3, GABARAPL1, GABARAPL2 and GABARAP, to reveal a C-terminal glycine. Exposure of the glycine at the C-terminus is essential for ATG8 proteins conjugation to phosphatidylethanolamine (PE) and insertion to membranes, which is necessary for autophagy. ATG4B is mainly expressed in the skeletal muscle, brain, heart, liver and pancreas.
Specifications

Specifications

ATG4b
Recombinant Polyclonal
0.5 mg/mL
PBS with 0.09% sodium azide
Q9Y4P1
AUTL1, APG4 autophagy 4 homolog B, AUT-like 1 cysteine endopeptidase, autophagin-1, autophagy-related cysteine endopeptidase 1, autophagy-related protein 4 homolog B, cysteine protease ATG4B, hAPG4B
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
23192
Immunocytochemistry, Immunofluorescence, Western Blot
1HCLC
Unconjugated
ATG4B
Liquid
ATG4B
full-length protein corresponding to Human ATG4b
Protein A
RUO
Antibody
Oligoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit