Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-ATG4B, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$181.00 - $399.00
Specifications
Antigen | ATG4B |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 2-4 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224735SS
|
Novus Biologicals
NBP224735SS |
0.025 mg |
Each for $181.00
|
|
|||||
NBP224735
|
Novus Biologicals
NBP224735 |
0.1 mg |
Each for $399.00
|
|
|||||
Description
ATG4B Polyclonal specifically detects ATG4B in Human, Mouse, Bovine samples. It is validated for Western Blot.Specifications
ATG4B | |
Western Blot 2-4 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Bovine | |
APG4 autophagy 4 homolog B, APG4 autophagy 4 homolog B (S. cerevisiae), APG4B, ATG4 autophagy related 4 homolog B (S. cerevisiae), AUTL1MGC1353, AUT-like 1 cysteine endopeptidase, Autophagin-1, Autophagy-related cysteine endopeptidase 1, Autophagy-related protein 4 homolog B, cysteine protease ATG4B, DKFZp586D1822, EC 3.4.22, EC 3.4.22.-, hAPG4B, KIAA0943 | |
ATG4B | |
IgG | |
Protein G purified | |
The amino acid sequence used as immunogen is 100% homologous in human, mouse and rat, 80% homologous in chicken and 75% homologous in pufferfish, xenopus and zebrafish. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Autophagy, Cancer | |
0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
23192 | |
A portion of amino acid 1-50 of human APG4B was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title