Learn More
ATG4C Sheep anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen OSA00013G
Catalog No. PIOSA00013G
Add to cart
Description
Description
Reconstitute with 500 µL of distilled water. Centrifuge to remove any insoluble material.
Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some maligt tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed.
Specifications
Specifications
ATG4C
Polyclonal
PBS with no preservative
Q96DT6, Q811C2
ATG4C, autophagin-3, APG4C, AUTL1, AUTL3, APG4-C, FLJ14867, ATG4 autophagy related 4 homolog C (S. cerevisiae), Autophagy-related protein 4 homolog C,
Sheep
IgG
500 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
242557, 313391, 84938
Immunohistochemistry, Western Blot
Unconjugated
ATG4C
Lyophilized
ATG4C
A synthetic peptide from human cysteine protease ATG4C
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human, Mouse, Rat