Learn More
ATG4C Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA118294
Catalog No. 01-670-547
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-18294 detects ATG4C from human and rat samples. PA1-18294 has been successfully used in immunohistochemistry, immunofluorescence, and Western blot procedures. The PA1-18294 immunogen is a synthetic peptide corresponding to the C-terminal residues of human ATG4C, conjugated to Blue Carrier Protein. The sequence is 100% conserved in mouse and rat. Reconstitute with 100 μL of distilled water.
Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some maligt tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases. Alternate transcriptional splice variants, encoding the same protein, have been characterized.
Specifications
Specifications
ATG4C
Polyclonal
whole serum with no preservative
Q96DT6
Cysteine protease ATG4C, Autophagy-related protein 4 homolog C, Autophagin-3, Autophagy-related cysteine endopeptidase 3, AUT-like 3 cysteine endopept
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
ATG4C
Lyophilized
ATG4C
Synthetic peptide corresponding to the C-terminal residues of human ATG4C.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84938