Learn More
ATG4D Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSA00035W
Catalog No. PIOSA00035W
Add to cart
Description
Description
Reconstitute with 150 µL of distilled water.
Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some maligt tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases.
Specifications
Specifications
ATG4D
Polyclonal
whole serum with no preservative
Q86TL0, Q8BGV9
Cysteine protease ATG4D, autophagin-4, APG4-D, APG4D, AUTL4
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry, Western Blot
Unconjugated
ATG4D
Lyophilized
ATG4D
A synthetic peptide from human, mouse cysteine protease ATG4D
150 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
235040, 84971