Learn More
ATG5 Rabbit anti-Bovine, Human, Mouse, Non-human primate, Porcine, Rat, Xenopus laevis, Zebrafish, HRP, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA522857
Catalog No. PIPA522857
Add to cart
Description
Description
In Western Blot, a band is seen ~56 kDa representing the ATG5-ATG12 complex, the molecular weight of human ATG5 is ~33 kDa. Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended. Suggested positive control: mouse wildtype ES cell lysate.
ATG5 (Autophagy Related 5) is an important element for autophagy and may play an important role in the apoptotic process. ATG5 is also involved in other cellular processes that include mitochondrial quality control after oxidative damage, negative regulation of the innate anti-viral immune response, lymphocyte development and proliferation, MHC II antigen presentation, and adipocyte differentiation. Following conjugation to ATG12, the conjugate participates in the formation of autophagosome. ATG5 contributes to autophagic cell death by interacting with Fas-associated protein with death domain (FADD). The ATG5-ATG12 conjugate forms a cup-shaped isolation membrane that then detaches from the membrane immediately before or after autophagosome formation is completed. APG5 may play a role in the apoptotic process, possibly within the modified cytoskeleton. Further, APG5 expression is a relatively late event in the apoptotic process, occurring downstream of caspase activity. The APG5-APG12 conjugate also associates with innate immune response proteins such as RIG-I and VISA (also known as IPS-1), inhibiting type I interferon production and permitting viral replication in host cells. Diseases associated with ATG5 dysfunction include spinocerebellar ataxia.
Specifications
Specifications
ATG5
Polyclonal
HRP
ATG5
Liquid
atg5, atg5.L
A synthetic peptide made to an N-terminal region (within residues 1-50) of the human ATG5 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Human, Mouse, Non-human primate, Porcine, Rat, Xenopus laevis, Zebrafish
Immunohistochemistry, Western Blot
0.92 mg/mL
PBS with no preservative
Q3MQ24, Q9H1Y0, Q99J83, Q3MQ04, Q3MQ06
APG5, APG5-LIKE, APG5L, ASP, hAPG5, apoptosis specific protein, apoptosis-specific protein, autophagy protein 5
Rabbit
IgG
100 μL
4° C, store in dark
Primary
11793, 365601, 443756, 494180, 532686, 641362, 9474