Learn More
ATG7 Sheep anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen OSA00008W
Catalog No. PIOSA00008W
Add to cart
Description
Description
Reconstitute with 150 µL of distilled water. Centrifuge to remove any insoluble material.
This gene was identified based on homology to Pichia pastoris GSA7 and Saccharomyces cerevisiae APG7. In the yeast, the protein appears to be required for fusion of peroxisomal and vaculuolar membranes. The protein shows homlogy to the ATP-binding and catalytic sites of the E1 ubiquitin activating enzymes. Functions as an E1 enzyme essential for multisubstrates such as GABARAPL1 and ATG12. Forms intermediate conjugates with GABARAPL1 (GABARAPL2, GABARAP or MAP1ALC3). Formation of the final GABARAPL1-PE conjugate is essential for autophagy. Interacts with ATG3 and ATG12. The complex, composed of ATG3 and ATG7, plays a role in the conjugation of ATG12 to ATG5. Widely expressed, especially in kidney, liver, lymph nodes and bone marrow. The C-terminal part of the protein is essential for the dimerization and interaction with ATG3 and ATG12.
Specifications
Specifications
ATG7
Polyclonal
whole serum with no preservative
O95352, Q9D906, Q641Y5
ATG7, APG7-like, APG7L, hAGP7, GSA7, APG7L, DKFZp434N0735, ubiquitin-activating enzyme E1-like protein
Sheep
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry
Unconjugated
ATG7
Lyophilized
ATG7
A synthetic peptide c-terminal region of human autophagy-related protein 7
150 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10533, 312647, 74244