Learn More
ATG9B Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSA00044W
Catalog No. PIOSA00044W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 150 μL of distilled water.
Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1. ATG9B plays a role in autophagy and it′s highly expressed in placenta and pituitary gland.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ATG9B
Polyclonal
Atg9b
Autophagy-related protein 9B, APG9L2, NOS3AS, SONE
Rabbit
150 μL
Primary
Mouse, Rat
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
Q6EBV9
ATG9B
A synthetic peptide from n-terminal region of mouse autophagy-related protein 9B
RUO
213948, 499973
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Recommendations
Recommendations