Learn More
ATGL Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA116974
Catalog No. 01-670-151
Add to cart
Description
Description
Suggested positive control: human adipose tissue lysates and mouse 3T3-L1 cell lysates, antigen standard for PNPLA2 (transient overexpression lysate).
Lipolytic enzymes are required for mobilization of fatty acids from triglyceride stores in adipose tissue. Energy homeostasis is affected by dysfunctional lipolysis and may contribute to the pathogenesis of obesity and insulin resistance. Until recently, hormone-sensitive lipase (HSL) was the only enzyme known to hydrolyze triglycerides in mammalian adipose tissue. It is now thought that a second enzyme, adipose triglyceride lipase (ATGL), catalyzes the initial step in triglyceride hydrolysis. ATGL is highly expressed in adipose tissue of mice and humans. It exhibits high substrate specificity for triacylglycerol and is associated with lipid droplets. Inhibition of ATGL markedly decreases total adipose acyl-hydrolase activity. Thus, ATGL and HSL coordinately catabolize stored triglycerides in adipose tissue of mammals.
Specifications
Specifications
ATGL
Polyclonal
Unconjugated
PNPLA2
Liquid
PNPLA2
Synthetic peptide corresponding to residues 400-500 of human ATGL.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
tris glycine with 150mM NaCl and 0.05% sodium azide
Q96AD5, Q8BJ56
Adipose triglyceride lipase antibody, transport secretion protein (TTS) antibody, desnutrin, calcium-independent phospholipase A2z (iPL-A2z) antibody,
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57104, 66853