Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ATP synthase C mature, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131242

Catalog No. NBP131242


Add to cart

Description

Description

ATP synthase C mature Polyclonal antibody specifically detects ATP synthase C mature in Human samples. It is validated for Western Blot.
Specifications

Specifications

ATP synthase C mature
1.0mg/mL
Western Blot 1:500-1:3000
Affinity Purified
ATP5MC1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human ATP5G1. The exact sequence is proprietary.
14 kDa
100 ul
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Expected to cross react based on sequence identity: Mouse (80%), Sheep (86%), Rhesus Monkey (93%), Bovine (85%). Not yet tested in any other species.
Western Blot
Unconjugated
PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal
ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C1 (subunit 9), ATP5A, ATP5G, ATPase protein 9, ATPase subunit 9, H+ transporting, mitochondrial F0 complex, subunit C1 (subunit 9), isoform 1, mitochondrial, mitochondrial ATP synthase, subunit 9, mitochondrial ATP synthase, subunit C
Rabbit
IgG
Antigen Affinity-purified
RUO
Primary
516
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only