Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP Synthase β Mouse, Unlabeled, Clone: 10, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612519

 View more versions of this product

Catalog No. BDB612519


Add to cart

Description

Description

ATP synthase is a large enzyme complex that uses an electrochemical H+ or Na+ gradient to synthesize ATP from ADP and Pi, providing the organism with the ATP needed for energy. The complex consists of two major units, F0 and F1. F0 is embedded in the inner membrane of the mitochondria and, due to its hydrophobic nature, translocates protons across this membrane. F1 is the catalytic portion in the matrix region of the mitochondria and is comprised of α, β, γ, δ, and ε subunits at a 3:3:1:1:1 ratio. The β subunit is synthesized in the nuclear genome,transported to the mitochondria, and assembled with the other subunits. It is encoded by a single copy gene, is ubiquitously expressed and highly conserved among species. ATP synthase β contains an Ets domain binding site, which is a main site for promoter activity. Ets proteins contain domains that are involved in transcriptional activation, protein-protein interactions, and intramolecular repression of DNA binding. This site acts as a target for transcriptional control by the Ets family of transcription factors. Thus, ATP synthase β is involved in the synthesis of ATP and is controlled in part by ETS family proteins.

Bioimaging, Immunofluorescence, Western Blotting

Specifications

Specifications

ATP Synthase β
10
Unconjugated
Affinity Purified
Human ATP Synthase β aa. 428-539
150μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.