Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

ATP Synthase β Mouse, Unlabeled, Clone: 10, BD

Mouse Monoclonal Antibody

$198.00 - $350.00

Specifications

Antigen ATP Synthase β
Isotype IgG1
Immunogen Human ATP Synthase β aa. 428-539
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB612518
View Documents
BD Biosciences
612518
50μg Each for $198.00
N/A
BDB612519
View Documents
BD Biosciences
612519
150μg Each for $350.00
N/A
Description

Description

ATP synthase is a large enzyme complex that uses an electrochemical H+ or Na+ gradient to synthesize ATP from ADP and Pi, providing the organism with the ATP needed for energy. The complex consists of two major units, F0 and F1. F0 is embedded in the inner membrane of the mitochondria and, due to its hydrophobic nature, translocates protons across this membrane. F1 is the catalytic portion in the matrix region of the mitochondria and is comprised of α, β, γ, δ, and ε subunits at a 3:3:1:1:1 ratio. The β subunit is synthesized in the nuclear genome,transported to the mitochondria, and assembled with the other subunits. It is encoded by a single copy gene, is ubiquitously expressed and highly conserved among species. ATP synthase β contains an Ets domain binding site, which is a main site for promoter activity. Ets proteins contain domains that are involved in transcriptional activation, protein-protein interactions, and intramolecular repression of DNA binding. This site acts as a target for transcriptional control by the Ets family of transcription factors. Thus, ATP synthase β is involved in the synthesis of ATP and is controlled in part by ETS family proteins.

Bioimaging, Immunofluorescence, Western Blotting

Specifications

Specifications

ATP Synthase β
Human ATP Synthase β aa. 428-539
Unconjugated
Cell Biology
Murine
Affinity Purified
Monoclonal
Store undiluted at -20°C.
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
10
250μg/mL

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit