Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP1B1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525760

Catalog No. PIPA525760

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Na,K-ATPase is an integral membrane protein complex that hydrolyzes ATP to maintain the transmembrane gradients of Na+ and K+ found in most mammalian cells. The enzyme is comprised of an alpha and Beta subunit. The á-polypeptide has been shown to be the catalytically active subunit, whereas the Beta-polypeptide is glycosylated and appears to be necessary for the assembly and transport of the sodium pump to the plasma membrane. Na,K-ATPase Beta-1 subunit is expressed, exclusively, in epithelial cells.
Specifications

Specifications

ATP1B1
Polyclonal
PBS with 0.09% sodium azide
P05026
ATP1B, Beta 1-subunit of Na(+),K(+)-ATPase, Na, K-ATPase beta-1 polypeptide, adenosinetriphosphatase, sodium/potassium-dependent ATPase beta-1 subunit, sodium/potassium-transporting ATPase beta-1 chain, sodium/potassium-transporting ATPase subunit beta-1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
481
Immunohistochemistry, Western Blot
Unconjugated
ATP1B1
Liquid
ATP1B1
KLH conjugated synthetic peptide between 112-138 amino acids from the central region of human ATP1B1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit