Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ATP5E, DyLight 488, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP142805G

Catalog No. NBP142805G


Add to cart

Description

Description

ATP5E Polyclonal antibody specifically detects ATP5E in Human, Mouse, Rat, Porcine, Bovine, Xenopus, Drosophila (Negative) samples. It is validated for Western Blot.
Specifications

Specifications

ATP5E
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
ATP5E
Synthetic peptide made to a sequence between amino acids 2-51 of human ATP5E. [Swiss-Prot# P56381]
5 kDa
0.1 ml
Store at 4C in the dark.
Polyclonal
Human, mouse, rat, porcine, bovine and Xenopus. Does not react with Drosophila.
Western Blot
Western Blot
P56381
ATP synthase, H+ transporting, mitochondrial F1 complex, epsilon subunit, ATPE, F(0)F(1)-ATPase, H(+)-transporting two-sector ATPase, MGC104243, mitochondrial, mitochondrial ATP synthase epsilon chain, mitochondrial ATPase
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
514
Bovine, Drosophila (Negative), Human, Mouse, Porcine, Rat, Xenopus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only