Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP5E Rabbit anti-Bovine, Human, Mouse, Porcine, Rat, Xenopus laevis, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523007

Catalog No. PIPA523007


Add to cart

Description

Description

This antibody does not react with drosophila. Suggested positive control: HEK293 cell lysate.

ATP5E encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). ATP5E encodes the epsilon subunit of the catalytic core. Two pseudogenes of this gene are located on chromosomes 4 and 13. [provided by RefSeq].
Specifications

Specifications

ATP5E
Polyclonal
Unconjugated
ATP5E
Liquid
atp5d.L, ATP5E
Synthetic peptide made to an N-terminal portion of human ATP5E (within residues 1-50).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Human, Mouse, Porcine, Rat, Xenopus laevis
Western Blot
1 mg/mL
PBS with 0.1% sodium azide
P05632, P56381, P56382, P29418
ATPE, MC5DN3, ATP synthase subunit epsilon, mitochondrial, F(0)F(1)-ATPase, H(+)-transporting two-sector ATPase, mitochondrial ATP synthase epsilon chain, mitochondrial ATPase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100134963, 245958, 447106, 514, 617230, 67126
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit